


N,N-Dimethylformamide dimethyl acetal ∼ 97%, AcroSeal®
Catalog # 446151000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3980 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:119.16 g/mol
Boiling Pt:102…104 °C (760 mmHg)
Density:0.89 g/cm³
Flash Pt:7 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS07
:GHS02
:GHS05
CAS Number:4637-24-5
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482