Order Entry
Italy
ContactUsLinkComponent
1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester 97%
 
1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester 97%
  ACRO445580010
 :  
undefined
 
1-[(2-Thienyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester 97%
  ACRO445580010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Off-white to yellow Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    54 to 59 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

 :
 :57…59 °C
 :290.19 g/mol
 :GHS07
 :Ambient
 :864754-19-8
 :C₁₄H₁₉BN₂O₂S
 :MFCD08706027