
Phenylboronic acid neopentyl glycol ester 97%
Catalog # ACRO445500050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White Solid
- Infrared spectrum:Conforms
- Melting point:62 to 67 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:190.05 g/mol
Melting Pt:65…67 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5123-13-7
Formula:C₁₁H₁₅BO₂
MDL Number:MFCD05663851