
2-Aminophenylboronic acid hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:863753-30-4
:
Formula:C₆H₉BClNO₂
MDL Number:MFCD02258096
:173.4052 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder or solid
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together