Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
 
Boron trifluoride - methanol complex 12% 1.5 M in methanol, AcroSeal®
 
undefined
 
 
 
 
Boron trifluoride - methanol complex 12% 1.5 M in methanol, AcroSeal®

 

  • Appearance:
    Colorless to amber Solution
  • Assay:
    10 to 15 % w/v (BF3)

 

Frequently Bought Together