

Acetone, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Boiling Pt:56 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.79 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
UN:1090
ADR:3,II
:GHS07
:GHS02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3580 to 1.3600 (20°C, 589 nm)
- Density:0.790 to 0.792 kg/l
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together