Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Phenazopyridine Hydrochloride, 99%, Analytical Standard
Phenazopyridine Hydrochloride, 99%, Analytical Standard
Phenazopyridine Hydrochloride, 99%, Analytical Standard
ACRO441921000
:
Thermo Fisher Scientific
:
Phenazopyridine Hydrochloride, 99%, Analytical Standard
ACRO441921000
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle
Pk:
100 mg
CAS No.:
136-40-3
Description:
Phenazopyridine hydrochloride, 99%
Formula:
C₁₁H₁₁N₅.HCl
Description:
Phenazopyridine hydrochloride