
3-Bromo-5-formylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113118-81-3
:
Formula:C₆H₄BrNO
MDL Number:MFCD03265758
:186.01 g/mol
Melting Pt:96…98 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:95 to 99 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together