
3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one 97%
Catalog # ACRO440990010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- NMR (1H-NMR):Consistent with structure
- Elemental Analysis:73.24 to 79.34 % (C)
Specification Test Results
About this item
:
:346.366 g/mol
Melting Pt:243…246 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:34932-07-5
Formula:C₂₂H₁₉O₂P
MDL Number:MFCD00065318