
6,7-Dimethoxy-1-tetralone 97%
:13575-75-2
:
:C₁₂H₁₄O₃
:MFCD00134100
:206.24 g/mol
:95…98 °C
:GHS07
:Ambient
- Appearance:White to off-white to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:95 to 100 °C
- HPLC:≥96.0 %
Frequently Bought Together