Order Entry
Italy
ContactUsLinkComponent
2-Pyridineboronic acid-N-phenyldiethanolamine ester 50 - 70%
Flammable
Irritant
2-Pyridineboronic acid-N-phenyldiethanolamine ester 50 - 70%
Certificates
undefined
Flammable
Irritant
2-Pyridineboronic acid-N-phenyldiethanolamine ester 50 - 70%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Additional info:
    Contains varying amounts of IPA and N-phenyldiethanolamine
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together