Order Entry
Italy
ContactUsLinkComponent
 
Poly(methylhydrosiloxane) trimethylsilyl terminated
 
undefined
Poly(methylhydrosiloxane) trimethylsilyl terminated

 

  • Appearance:
    Colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3960 to 1.4000 (20°C, 589 nm)
  • Viscosity:
    15 to 25 cSt

 

 

Frequently Bought Together