Order Entry
Italy
Orders LinkContactUsLinkComponent
Adipic semialdehyde methyl ester 95%

Adipic semialdehyde methyl ester 95%

Glass bottle, 250 mg
Codice catalogo ACRO438912500
Irritant
Nome del fornitore:   Thermo Fisher Scientific
Adipic semialdehyde methyl ester 95%
Codice catalogo ACRO438912500
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  6654-36-0
Adipic semialdehyde methyl ester 95%
Adipic semialdehyde methyl ester 95%

Glass bottle, 250 mg

Numero CAS:  6654-36-0
Price
140,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  6-Oxohexanoic Acid Methyl Ester, Methyl 6-oxohexanoate, Hexanoic acid,6-oxo-,methyl ester
  • CAS Number:6654-36-0
  • Formula:C₇H₁₂O₃
  • MDL Number:MFCD00063422
  • Molecular Weight:144.17 g/mol
  • Boiling Pt:59…61 °C (1Torr)
  • Storage Temperature:Freezer

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless Liquid
  • Infrared spectrum:
    Conforms
  • NMR (1H-NMR):
    Consistent with structure

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg

Specifications