Order Entry
Italy
ContactUsLinkComponent
2-Methyl-2-phenylpropylmagnesium chloride 0.5 M in diethyl ether, AcroSeal®
Flammable
Corrosive
Irritant
2-Methyl-2-phenylpropylmagnesium chloride 0.5 M in diethyl ether, AcroSeal®
Catalog # 438751000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
2-Methyl-2-phenylpropylmagnesium chloride 0.5 M in diethyl ether, AcroSeal®
Catalog # 438751000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to yellow to grey Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    0.40 to 0.60 M

Specification Test Results

About this item

 :
Density:0.76 g/cm³
Flash Pt:–28 °C
ADR:3,I
 :192.97 g/mol
UN:2924
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:35293-35-7
Formula:C₁₀H₁₃ClMg
MDL Number:MFCD00075586