Order Entry
Italy
ContactUsLinkComponent
2,3-Dimethyl-2-butene 1 M in THF, AcroSeal®
Flammable
Irritant
Health
2,3-Dimethyl-2-butene 1 M in THF, AcroSeal®
Catalog # 438721000.
CAS Number:  
undefined
Flammable
Irritant
Health
2,3-Dimethyl-2-butene 1 M in THF, AcroSeal®
Catalog # 438721000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    0.95 to 1.10 M

Specification Test Results

About this item

 :
 :84.16 g/mol
Boiling Pt:73 °C (1013 hPa)
Melting Pt:–75 °C
Density:0.857 g/cm³
Flash Pt:–21 °C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:563-79-1
Formula:C₆H₁₂
MDL Number:MFCD00008897