Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
Irritant
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
Catalog # ACRO438020010
CAS Number:  
undefined
Irritant
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
Catalog # ACRO438020010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Off-white to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :255.336 g/mol
Melting Pt:223.5…224.5 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:365996-05-0
Formula:C₁₁H₁₇N₃O₂S
MDL Number:MFCD08448157