Order Entry
Italy
ContactUsLinkComponent
 
 
5-Bromo-2-methylindole 98%
 
undefined
 
5-Bromo-2-methylindole 98%

 

  • Appearance:
    Yellow to pink to tan Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    103 to 107 °C
  • HPLC:
    ≥97.5 %

 

 

Frequently Bought Together