
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate 97%
Catalog # ACRO437520100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to light yellow Powder or crystalline powder
- Infrared spectrum:Conforms
- Melting point:73 to 77 °C
- GC:≥96.0 %
Specification Test Results
About this item
CAS Number:157634-00-9
:
Formula:C₁₁H₂₁NO₃
MDL Number:MFCD04114966
:215.293 g/mol
Melting Pt:74.4…75.7 °C
:GHS07
Storage Temperature:Ambient