Order Entry
Italy
ContactUsLinkComponent
3,3',5-Triiodo-L-thyronine 95%
Irritant
3,3',5-Triiodo-L-thyronine 95%
Certificates
undefined
Irritant
3,3',5-Triiodo-L-thyronine 95%

Specification Test Results

  • Appearance:
    White to cream to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥94 %
  • Specific optical rotation:
    +18° to +28° (20°C, 589 nm) (c=2,ethanol/0.5N HCl(4:1))

Specification Test Results

Related Information

Frequently Bought Together