
1-Boc-4-(aminomethyl)piperidine 97%
Catalog # ACRO436610050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
CAS Number:144222-22-0
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD01076207
:214.31 g/mol
Boiling Pt:237…238 °C (760 mmHg)
Density:1.013 g/cm³
Flash Pt:>100 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05