



Pentadecylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 436160500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.45 to 0.55 M
- Melting point:80 to 83 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:315.62 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:78887-70-4
Formula:C₁₅H₃₁BrMg
MDL Number:MFCD00209607