
Nortropinone hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25602-68-0
:
Formula:C₇H₁₂ClNO
MDL Number:MFCD03613582
:161.63 g/mol
Melting Pt:194.7…195.8 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white to light brown Powder or solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 to ≤104.0 %
- GC:≥98.0 %
- Specific optical rotation:+12° to +15° (20°C, 589 nm) (Neat)
Specification Test Results
Frequently Bought Together