


Pentylmagnesium bromide 2 M in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:693-25-4
:
Formula:C₅H₁₁BrMg
MDL Number:MFCD00000045
Density:0.947 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
:175.35 g/mol
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Yellow to brown to black Solution
- Titration Grignard:1.90 to 2.20 M
- Refractive index:1.5160 to 1.5180 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Related Information
Frequently Bought Together