


4-Ethoxy-1,1,1-trifluorobut-3-en-2-one 97%
Catalog # ACRO435360050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Stabilizer:0.5% BHT
Specification Test Results
About this item
:
:168.115 g/mol
Boiling Pt:51…53 °C (12 mmHg)
Density:1.18 g/cm³
Flash Pt:51 °C
Storage Temperature:Refrigerator
UN:2929
ADR:6.1,II
:GHS02
:GHS06
:GHS08
CAS Number:17129-06-5
:MFCD08461613
Formula:C₆H₇F₃O₂
MDL Number:MFCD00192131