Order Entry
Italy
ContactUsLinkComponent
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Irritant
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Catalog # ACRO435235000
CAS Number:  
undefined
Irritant
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Catalog # ACRO435235000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Orange to red to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • NMR (1H-NMR):
    Consistent with structure
  • NMR (19F-NMR):
    Consistent with structure
  • NMR (31P-NMR):
    Consistent with structure
  • Specific optical rotation:
    +35° to +45° (20°C, 589 nm) (c=1 in methanol)

Specification Test Results

About this item

 :
 :554.19 g/mol
Melting Pt:152…156 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:213343-65-8
Formula:C₂₂H₃₈BF₄P₂Rh
MDL Number:MFCD09038435