
2-Chloro-4-methoxypyridine 97%
Catalog # ACRO434930010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:143.57 g/mol
Boiling Pt:229…230 °C (760 mmHg)
Density:1,258 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:17228-69-2
Formula:C₆H₆ClNO
MDL Number:MFCD02093951