Order Entry
Italy
ContactUsLinkComponent
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine 97%
Irritant
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine 97%
Catalog # ACRO434092500
CAS Number:  
undefined
Irritant
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine 97%
Catalog # ACRO434092500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • NMR:
    ≥96.0 %

Specification Test Results

About this item

 :
Melting Pt:87…93 °C
 :381.459 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1091606-67-5
Formula:C₂₆H₂₄NP
MDL Number:MFCD11045444