
4-Vinylphenol (95%) 10% in propylene glycol
ACRO434040250
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Appearance:Clear colorless to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4420 to 1.4520 (20°C, 589 nm)
- Assay:9 to 15 %
- Density:(20°C) 1.038 to 1.048 g/ml
:2628-17-3
:
:C₈H₈O
:MFCD00017593
:120.151 g/mol
:189 °C (760 mmHg)
:1.039 g/cm³
:GHS08
:Refrigerator