Order Entry
Italy
ContactUsLinkComponent
3-Thienylmagnesium iodide 0.3 M in THF, AcroSeal®
Flammable
Irritant
Corrosive
Health
3-Thienylmagnesium iodide 0.3 M in THF, AcroSeal®
Catalog # 433650500.
CAS Number:  
undefined
Flammable
Irritant
Corrosive
Health
3-Thienylmagnesium iodide 0.3 M in THF, AcroSeal®
Catalog # 433650500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Solution
  • Titration Grignard:
    0.20 to 0.40 M
  • Water:
    ≤0.005 %
  • GC:
    ≥98.0 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

About this item

 :
 :234.337 g/mol
Boiling Pt:65 °C (760 mmHg)
Density:0.904 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS02
 :GHS07
 :GHS05
 :GHS08
CAS Number:172035-86-8
Formula:C₄H₃IMgS
MDL Number:MFCD03427252