
Ethyl-2-hydrazinylacetate hydrochloride 97%
Catalog # ACRO433150100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Light yellow to light orange Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:146 to 151 °C
- Titration Argentometric:≥96.0 to ≤104.0 %
Specification Test Results
About this item
:
:154.594 g/mol
Melting Pt:147…149 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6945-92-2
Formula:C₄H₁₁ClN₂O₂
MDL Number:MFCD00012923