

2-(Trifluoromethyl)pyrazine 97%
Catalog # ACRO432610050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
:148.09 g/mol
Boiling Pt:116…118 °C (760 mmHg)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:61655-67-2
Formula:C₅H₃F₃N₂
MDL Number:MFCD06797734