Order Entry
Italy
ContactUsLinkComponent
4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane 90%
Irritant
Flammable
4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane 90%
Catalog # ACRO430450050
CAS Number:  
undefined
Irritant
Flammable
4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane 90%
Catalog # ACRO430450050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to brown Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥88.0 %
  • Stabilizer:
    Phenothiazine stabilizer

Specification Test Results

About this item

 :
 :168.04 g/mol
Boiling Pt:47 °C (9mbar)
Density:0.88 g/cm³
Flash Pt:42 °C
UN:1993
ADR:3,III
 :GHS07
 :GHS02
Storage Temperature:Freezer
CAS Number:126726-62-3
Formula:C₉H₁₇BO₂
MDL Number:MFCD08276843