Order Entry
Italy
ContactUsLinkComponent
L-Adrenaline bitartrate 98+%
Toxic
L-Adrenaline bitartrate 98+%
Certificates
undefined
Toxic
L-Adrenaline bitartrate 98+%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    147 to 152 °C (Dec)
  • Loss on drying:
    ≤0.5 %
  • Specific optical rotation:
    -20° to -24° (20 °C, 589 nm) (c=4,1M HCl)
  • Titration with HClO4:
    ≥98.0 %
  • Insoluble matter:
    ≤0.005 % (in water)

Specification Test Results

Related Information

Frequently Bought Together