
1-Methyl-1H-indazole-4-carboxaldehyde 95%
:1053655-56-3
:
:C₉H₈N₂O
:MFCD10699167
:160.18 g/mol
:77.5…79 °C
:GHS07
:Ambient
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:76 to 80 °C
- HPLC:≥94.0 %
- Sulfate (SO4):≤0.05 %
- Ammonium (NH4):≤0.01 %
- Arsenic (As):≤1 ppm
- Copper (Cu):≤0.002 %
- Iron (Fe):≤0.005 %
- Lead (Pb):≤0.002 %
- Nickel (Ni):≤0.002 %
- Insoluble matter:≤0.025 % (in water and ppt by NH4OH)
Frequently Bought Together