


Bromine 1 M in trimethyl phosphate, AcroSeal®
Catalog # 428721000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to red to brown Solution
- Titration with Na2S2O3:0.9 to 1.5 M
- Sulfated ash:≤0.1 % (1 g)
- Appearance of solution:(5 g in 50 ml water) clear and not more intensely colored than ref. soln. Y6
- Identification A:Passes test
- Identification B:Passes test
- Specific optical rotation:+12.0° to +12.8° (20°C, 589 nm) (c=20, H2O) (on dried substance)
- Titration with NaOH:99.5 to 101.0 % (on dried substance)
- Sulfates:≤150 ppm
- Chlorides:≤100 ppm
- Calcium (Ca):≤200 ppm
- Impurity:≤350 ppm (as anhydrous oxalic acid)
Specification Test Results
About this item
:
:159.81 g/mol
Density:1.307 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
UN:1744
ADR:8,I
:GHS05
:GHS06
:GHS08
CAS Number:7726-95-6
Formula:Br₂
MDL Number:MFCD00010896