N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 98%
Codice catalogo ACRO428640010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
- Specific optical rotation:+42° to +48° (20°C, 589 nm) (c=1 in DMF)
Specification Test Results
About this item
:
:434.881 g/mol
Melting Pt:223 °C
Storage Temperature:Ambient
CAS Number:21653-40-7
Formula:C₁₉H₂₃ClN₆O₄
MDL Number:MFCD00063682