
Methyl-1-hydroxy-2-naphthoate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:948-03-8
:
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00043823
:202.21 g/mol
Melting Pt:76…80 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:74 to 80 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together