Order Entry
Italy
ContactUsLinkComponent
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 97%
Irritant
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 97%
Certificates
undefined
Irritant
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14) 97%
Specification Test Results
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    60.57 to 64.96 % C