Order Entry
Italy
ContactUsLinkComponent
Isobutanol for HPLC
Irritant
Corrosive
Flammable
Isobutanol for HPLC
Catalog # ACRO412640010
   Safer Human and Environmental Health
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Isobutanol for HPLC
Catalog # ACRO412640010
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.45 %
  • UV:
    at 219 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %

Specification Test Results

About this item

 :
 :74.12 g/mol
Boiling Pt:108 °C (760 mmHg)
Melting Pt:–108 °C
Density:0.8 g/cm³
Flash Pt:28 °C
UN:1212
ADR:3,III
 :GHS07
 :GHS05
 :GHS02
Storage Temperature:Ambient
CAS Number:78-83-1
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740