
1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207857-15-6
:
Formula:C₁₂H₂₀F₃N₃O₆S
MDL Number:MFCD02683528
:391.36 g/mol
Melting Pt:111…113 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Solid
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together