Order Entry
Italy
ContactUsLinkComponent
 
 
4,5-Dichloro-2-methyl-2H-pyridazin-3-one 97%
 
undefined
 
4,5-Dichloro-2-methyl-2H-pyridazin-3-one 97%

 

  • Appearance:
    Off-white to yellow to brown Powder or crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    87 to 92 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together