
5-Bromo-2-fluorobenzoic acid 97%
Catalog # ACRO398590050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:150 to 154 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:219.01 g/mol
Melting Pt:141…145 °C
ADR:6.1,III
:GHS07
Storage Temperature:Ambient
CAS Number:146328-85-0
Formula:C₇H₄BrFO₂
MDL Number:MFCD00143423