Order Entry
Italy
ContactUsLinkComponent
3-Bromo-4-formylpyridine 97%
Irritant
3-Bromo-4-formylpyridine 97%
Certificates
undefined
Irritant
3-Bromo-4-formylpyridine 97%

Specification Test Results

  • Appearance:
    Light yellow to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    79 to 83 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together