N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 95%
:107819-90-9
:
:C₁₂H₂₂N₂O₄S
:MFCD00239356
:115…121 °C
:290.38 g/mol
:GHS07
:Ambient
- Appearance:White to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:118 to 125 °C
- HPLC:≥94 %
Frequently Bought Together