
4'-Iodoacetanilide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:622-50-4
:
Formula:C₈H₈INO
MDL Number:MFCD00016352
:261.0597 g/mol
Melting Pt:184…185 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Bleu-grey to purple Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together