

Tetramethylammonium triacetoxyborohydride 90+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109704-53-2
:
Formula:C₁₀H₂₂BNO₆
MDL Number:MFCD00012196
Melting Pt:95…98 °C
Storage Temperature:Ambient
UN:2813
ADR:4.3,I
:263.10 g/mol
:GHS02
:GHS07
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥90.0 %
Specification Test Results
Frequently Bought Together