
Titanium(IV) bromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-68-6
:
Formula:Br₄Ti
MDL Number:MFCD00011265
:367.5 g/mol
Boiling Pt:230 °C (760 mmHg)
Melting Pt:39 °C
Density:2,6 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Orange Powder and/or lumps or low melting solid
- Titration Argentometric:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together