Order Entry
Italy
ContactUsLinkComponent
 
 
4,5-Dichlorophthalic anhydride 98%
 
undefined
 
4,5-Dichlorophthalic anhydride 98%

 

  • Appearance:
    White to yellow to grey Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    185 to 189 °C
  • HPLC:
    ≥97.5 %

 

 

Frequently Bought Together