Methyl pelargonate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1731-84-6
:
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00009569
Boiling Pt:213 °C (760 mmHg)
Melting Pt:–30 °C
Density:0.874 g/cm³
Flash Pt:81 °C
:172.27 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4230 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together