Propyl propionate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-36-5
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009373
:116.16 g/mol
Boiling Pt:123 °C (760 mmHg)
Melting Pt:–76 °C
Density:0.881 g/cm³
Flash Pt:26 °C
Storage Temperature:Ambient
UN:3272
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3820 to 1.3940 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together